Learn More
Medchemexpress LLC HY-112451 5mg Medchemexpress, cFMS Receptor Inhibitor II CAS:959860-85-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1124515MG
Encompass_Preferred
Medchemexpress, HY-112451 5mg cFMS Receptor Inhibitor II CAS:959860-85-6 cFMS Receptor Inhibitor II is a CSF1R kinase inhibitor. CSF-1 is a cytokine [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2423