Learn More
Aat Bioquest CF-MU [6-Chloro-8-fluoro-umbelliferone] Fluorescence reference standard
SDP
Catalog No. 502979334
Encompass_Preferred
This compound is used as a calibration standard for CF-MU-based enzyme substrates.
Catalog No. 50-297-9334 Supplier Aat Bioquest Supplier No. 42
This compound is used as a calibration standard for CF-MU-based enzyme substrates.