Learn More
Aviva Systems Biology Corp CFP Antibody - middle region (ARP56382_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56382_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CFP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CFP. Alternate names: BFD, PFC, PFD, PROPERDINApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61836