Learn More
IW Tremont Co Inc LabExact CFP2 Fluted filter paper, 8um 18.5cm qualitative smooth bright white filters are neatly packaged in groups of 10, ready to use 100/pk
SDP
Supplier:  IW Tremont Co Inc LECFP2F185
Encompass_Preferred
LabExact Fluted Grade CFP2 filter paper is easier to use and more convenient than flat filters when used in a filter funnel. 8um retention with medium-slow flow rate. Very high loading capacity. Sharply creased, smooth bright white filters are neatly packaged in groups of 10, ready to use. Applications include general laboratory separation & clarification and soil analysis methods.
Catalog No. 50-223-7485