Learn More
IW Tremont Co Inc LabExact CFP41 cellulose filter paper, 25um 11cm very fast flow rates ideal for initial separation of gelatinous precipitates, extremely low ash content of <0.01%. Thickness 0.220mm 100/pk
SDP
Supplier:  IW Tremont Co Inc LECFP41110
Encompass_Preferred
LabExact Very fast flow rates and loose retention characteristics make Grade CFP41 ideal for initial separation of gelatinous precipitates. Retention of approximately 20 um. Extremely low ash content of <0.01%. Thickness 0.220mm.
Catalog No. 50-223-7540