Learn More
IW Tremont Co Inc LabExact CFP41 cellulose filter paper, 25um 15cm very fast flow rates ideal for initial separation of gelatinous precipitates, extremely low ash content of <0.01%. Thickness 0.220mm 100/pk
SDP
Catalog No. 502237542
Encompass_Preferred
LabExact Very fast flow rates and loose retention characteristics make Grade CFP41 ideal for initial separation of gelatinous precipitates. Retention of approximately 20 um. Extremely low ash content of <0.01%. Thickness 0.220mm.
This item is not returnable. View return policy
Catalog No. 50-223-7542 Supplier IW Tremont Co Inc Supplier No. LECFP41150
LabExact Very fast flow rates and loose retention characteristics make Grade CFP41 ideal for initial separation of gelatinous precipitates. Retention of approximately 20 um. Extremely low ash content of <0.01%. Thickness 0.220mm.