Learn More
IW Tremont Co Inc LabExact CFP42 cellulose filter paper, 2.5 um 12.5cm quantitative high retention rate ideal for any gravimetric analysis of very fine precipitates extremely low ash content of <0.01%. Thickness 0.170mm 100/pk
SDP
Supplier:  IW Tremont Co Inc LECFP42125
Encompass_Preferred
LabExact Grade CFP42 demonstrates an extremely high retention rate for a cellulose filter media. Its uniquely high retention rate makes it ideal for any gravimetric analysis of very fine precipitates. Retention of approximately 2.5 um. Extremely low ash content of <0.01%. Thickness 0.170mm.
Catalog No. 50-223-7549