Learn More
IW Tremont Co Inc LabExact CFP44 Cellulose filter paper, 3um 11.0cm quantitative, used in the analysis of samples requiring separation of very fine precipitates. extremely low ash content of <0.01%. Thickness 0.165mm 100/pk
SDP
Supplier:  IW Tremont Co Inc LECFP44110
Encompass_Preferred
LabExact Grade CFP44 is very similar to Grade CFP42 but demonstrating a slightly wider retention rate within a similar flow rate. Typically used in the analysis of samples requiring separation of very fine precipitates. . Retention of approximately 3 um. Extremely low ash content of <0.01%. Thickness 0.165mm.
Catalog No. 50-223-7562