Learn More
IW Tremont Co Inc LabExact CFP541 cellulose filter paper, 21-23um 15.0cm quantitative hardened with increased wet strength, medium flow rate extremely low ash content of <0.01% Thickness 0.20mm 100/pk
SDP
Catalog No. 502237574
Encompass_Preferred
LabExact Grade CFP541 quantitative cellulose filter media is ashless and hardened with increased wet strength. Fast flow rates and retention of approximately 21 - 23 um. Extremely low ash content of <0.01%. Thickness 0.200mm.
This item is not returnable. View return policy
Catalog No. 50-223-7574 Supplier IW Tremont Co Inc Supplier No. LECFP541150
LabExact Grade CFP541 quantitative cellulose filter media is ashless and hardened with increased wet strength. Fast flow rates and retention of approximately 21 - 23 um. Extremely low ash content of <0.01%. Thickness 0.200mm.