Learn More
GINKGO BIOWORKS INC (3P) Ginkgo Bioworks™ Cell Free Protein Synthesis - Ready96 (E. coli CFPS): 10 plates
Catalog No. NC3953812
Encompass
Ginkgo’s CFPS Ready96 consists of Ginkgo’s premier CFPS system, reformatted to meet high-throughput screening needs. Reactions are pre-mixed and aliquoted into a 96 well plate. The user must simply add DNA and water, and reactions will produce target protein within hours. The kit is an E. coli transcription/translation system optimized to produce proteins from a T7 RNA Polymerase promoter. Reactions consist of a bacterial cell lysate and a reagent mix that contain all components required for protein production. Each plate enables 96 reactions of 20 µL. Of this volume, 12 µL are pre-aliquotted, and the user supplies the remaining 8 µL of DNA, water, and any other desired reagents. Includes ten (10) plates. Storage at -80° C. For research use only.
Catalog No. NC3953812 Supplier GINKGO BIOWORKS INC (3P) Supplier No. CF103
Ginkgo’s CFPS Ready96 consists of Ginkgo’s premier CFPS system, reformatted to meet high-throughput screening needs. Reactions are pre-mixed and aliquoted into a 96 well plate. The user must simply add DNA and water, and reactions will produce target protein within hours. The kit is an E. coli transcription/translation system optimized to produce proteins from a T7 RNA Polymerase promoter. Reactions consist of a bacterial cell lysate and a reagent mix that contain all components required for protein production. Each plate enables 96 reactions of 20 µL. Of this volume, 12 µL are pre-aliquotted, and the user supplies the remaining 8 µL of DNA, water, and any other desired reagents. Includes ten (10) plates. Storage at -80° C. For research use only.