Learn More
Biochain Institute Inc cfPure MAX Cell-Free DNA Extraction Kit, 1 Kit (250 ml)/pk
SDP
Catalog No. 501871536
Encompass_Preferred
BioChain's cfPure Cell Free DNA Extraction Kit is designed for rapid and efficient purification of circulating cell free DNA (cfDNA). The system utilizes silica-coated paramagnetic particles to purify cell free DNA from less than 1 mL to greater than 10 mL of serum or plasma. The buffers were developed to ensure efficient recovery of 100 bp - 500 bp DNA fragments in order to maximize recovery of cell free DNA. The recovered DNA is suitable for a wide range of down-stream applications, such as bisulfite sequencing, NGS, and qPCR.
Catalog No. 50-187-1536 Supplier Biochain Institute Inc Supplier No. K5011625MA
BioChain's cfPure Cell Free DNA Extraction Kit is designed for rapid and efficient purification of circulating cell free DNA (cfDNA). The system utilizes silica-coated paramagnetic particles to purify cell free DNA from less than 1 mL to greater than 10 mL of serum or plasma. The buffers were developed to ensure efficient recovery of 100 bp - 500 bp DNA fragments in order to maximize recovery of cell free DNA. The recovered DNA is suitable for a wide range of down-stream applications, such as bisulfite sequencing, NGS, and qPCR.