Learn More
Beckman Coulter Sterile and Certified Free Tubes
Catalog No. NC2063764
Encompass_Preferred
The Beckman Coulter Sterile and Certified Free Tubes are sterile, certified DNAse, RNAse and pyrogen free polypropylene tubes available in 15mL and 50mL sizes. They have conical bottoms and leakproof, screw caps. The tubes are gamma irradiated and certified non-cytotoxic. They can be used for sample collection, preparation, storage and processing in molecular biology, tissue culture, microbiology, immunology, hematology, and other life science applications. The tubes provide reliable containment and are ideal for sensitive procedures requiring sterile technique.
Catalog No. NC2063764 Supplier Beckman Coulter Supplier No. C14299
The Beckman Coulter Sterile and Certified Free Tubes are sterile, certified DNAse, RNAse and pyrogen free polypropylene tubes available in 15mL and 50mL sizes. They have conical bottoms and leakproof, screw caps. The tubes are gamma irradiated and certified non-cytotoxic. They can be used for sample collection, preparation, storage and processing in molecular biology, tissue culture, microbiology, immunology, hematology, and other life science applications. The tubes provide reliable containment and are ideal for sensitive procedures requiring sterile technique.