Learn More
Selleck Chemical LLC CFSE 10mg 150347-59-4 Carboxyfluorescein succinimidyl ester
SDP
Supplier:  Selleck Chemical LLC S826910MG
Encompass_Preferred
Carboxyfluorescein succinimidyl ester (CFSE) is a fluorescent cell staining dye. CFSE is cell permeable and covalently couples, via its succinimidyl group, to intracellular molecules, notably, to intracellular lysine residues and other amine sources. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5868