Learn More
TARGETMOL CHEMICALS INC CFSE 25MG
SDP
Catalog No. 502576449
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Carboxyfluorescein succinimidyl ester (CFSE (CFDA-SE)) is a fluorescent cell staining dye. CFSE is cell permeable and covalently couples via its succinimidyl group to intracellular molecules notably to intracellular lysine residues and other amine sources. purity: 99%
Catalog No. 50-257-6449 Supplier TARGETMOL CHEMICALS INC Supplier No. T680225MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Carboxyfluorescein succinimidyl ester (CFSE (CFDA-SE)) is a fluorescent cell staining dye. CFSE is cell permeable and covalently couples via its succinimidyl group to intracellular molecules notably to intracellular lysine residues and other amine sources. purity: 99%