Learn More
Medchemexpress LLC HY-D0938 5mg Medchemexpress, CFSE CAS:150347-59-4 Purity:>98%
SDP
Catalog No. 501933214
Encompass_Preferred
Medchemexpress, HY-D0938 5mg CFSE CAS:150347-59-4 CFSE is a fluorescent dye which can track the cell division. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3214 Supplier Medchemexpress LLC Supplier No. HYD09385MG
Medchemexpress, HY-D0938 5mg CFSE CAS:150347-59-4 CFSE is a fluorescent dye which can track the cell division. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.