Learn More
enQuire Bioreagents CFTR Recombinant Monoclonal [Clone CFTR/1775R], Size: 100ug
Supplier:  enQuire Bioreagents 1080RBM5P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: CFTR/1775R. Clonality: Monoclonal. Host: Rabbit. Isotype: IgG kappa. Positive Control: MOLT-4 cells. Pancreas, Kidney or Placenta. Cellular Staining: Cell Surface and Cytoplasmic. Buffer: 10mM PBS with or without 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human CFTR fragment (aa258-385). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 165-170kDa.Chromosome: 7q31.2. Gene Symbol: CFTR. Gene ID: 1080. Ensemble: ENSG00000001626. Uniprot: NP_011514056.
Catalog No. 50-188-4587