Learn More
Medchemexpress LLC HY-111772 5mg Medchemexpress, CFTR corrector 1 CAS:2216712-66-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1117725MG
Encompass_Preferred
Medchemexpress, HY-111772 5mg CFTR corrector 1 CAS:2216712-66-0 CFTR corrector 1 (compound 1) is a modulator of cystic fibrosis transmembrane conductance regulator (CFTR) . CFTR corrector 1 (compound 1) facilitates the processing and trafficking of CFTR to increase the amount of CFTR at the cell surface [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2390