Learn More
Medchemexpress LLC HY-125381 5mg Medchemexpress, CFTR corrector 2 CAS:1628416-28-3 Purity:>98%
SDP
Catalog No. 501932605
Encompass_Preferred
Medchemexpress, HY-125381 5mg CFTR corrector 2 CAS:1628416-28-3 CFTR corrector 2 is a cystic fibrosis transmembrane conductance corrector (CFTR) , extracted from patent US20140274933 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2605 Supplier Medchemexpress LLC Supplier No. HY1253815MG
Medchemexpress, HY-125381 5mg CFTR corrector 2 CAS:1628416-28-3 CFTR corrector 2 is a cystic fibrosis transmembrane conductance corrector (CFTR) , extracted from patent US20140274933 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.