Learn More
TARGETMOL CHEMICALS INC CFTR CORRECTOR 4 5MG
SDP
Catalog No. 502581668
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CFTR corrector 4 is a potent and orally available transmembrane conductance regulator (CFTR) for cystic fibrosis and is a potent (RR) type active enantiomer. CFTR corrector 4 increases CFTR levels on the cell surface and is a potential compound for the study of cystic fibrosis. purity: 98%
Catalog No. 50-258-1668 Supplier TARGETMOL CHEMICALS INC Supplier No. T107765MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CFTR corrector 4 is a potent and orally available transmembrane conductance regulator (CFTR) for cystic fibrosis and is a potent (RR) type active enantiomer. CFTR corrector 4 increases CFTR levels on the cell surface and is a potential compound for the study of cystic fibrosis. purity: 98%