Learn More
Medchemexpress LLC CFTR(inh)-172 | 307510-92-5 | 99.8% | 409.40 | 1 ML
SDP
Catalog No. 5000392206
Encompass_Preferred
CFTR(inh)-172 is a potent and selective blocker of the CFTR chloride channel, reversibly inhibiting CFTR short-circuit current with a Ki of 300 nM. It is intended for research use only.
  • Potent and selective CFTR chloride channel blocker.
  • Reversibly inhibits CFTR short-circuit current in less than 2 minutes.
  • Nontoxic to FRT cells at concentrations up to 100 μM.
  • Reduces open probability by > 90% in CFTR channels.
  • Significantly reduces cholera toxin-induced fluid secretion in mouse models.
Catalog No. 50-003-92206 Supplier Medchemexpress LLC Supplier No. HY1667110MM/1ML
CFTR(inh)-172 is a potent and selective blocker of the CFTR chloride channel, reversibly inhibiting CFTR short-circuit current with a Ki of 300 nM. It is intended for research use only.
  • Potent and selective CFTR chloride channel blocker.
  • Reversibly inhibits CFTR short-circuit current in less than 2 minutes.
  • Nontoxic to FRT cells at concentrations up to 100 μM.
  • Reduces open probability by > 90% in CFTR channels.
  • Significantly reduces cholera toxin-induced fluid secretion in mouse models.