Learn More
Medchemexpress LLC HY-16671 50mg Medchemexpress, CFTR(inh)-172 CFTR Inhibitor-172;CFTRinh-172 CAS:307510-92-5 Purity:98%
SDP
Catalog No. 501638546
Encompass_Preferred
Medchemexpress,HY-16671 50mg CFTR(inh)-172 CFTR Inhibitor-172;CFTRinh-172 CAS:307510-92-5 Formula:C18H10F3NO3S2 CFTR(inh)-172 is a potent and selective blocker of the CFTR chloride channel; reversibly inhibited CFTR short-circuit current in less than 2 minutes with a Ki of 300 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8546 Supplier Medchemexpress LLC Supplier No. HY1667150MG
Medchemexpress,HY-16671 50mg CFTR(inh)-172 CFTR Inhibitor-172;CFTRinh-172 CAS:307510-92-5 Formula:C18H10F3NO3S2 CFTR(inh)-172 is a potent and selective blocker of the CFTR chloride channel; reversibly inhibited CFTR short-circuit current in less than 2 minutes with a Ki of 300 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.