Learn More
Selleck Chemical LLC CFTRinh-172 10mg 307510-92-5 CFTR inhibitor 172
SDP
Supplier:  Selleck Chemical LLC S713910MG
Encompass_Preferred
CFTRinh-172 (CFTR inhibitor 172) is a voltage-independent, selective CFTR inhibitor with Ki of 300 nM, showing no effects on MDR1, ATP-sensitive K+ channels, or a series of other transporters. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8025