Learn More
Medchemexpress LLC HY-16671 10mg , CFTR(inh)-172 CAS:307510-92-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1667110MG
Encompass_Preferred
HY-16671 10mg CFTR(inh)-172 CAS: 307510-92-5 CFTR(inh)-172 is a potent and selective blocker of the CFTR chloride channel; reversibly inhibits CFTR short-circuit current in less than 2 minutes with a Ki of 300 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0192