Learn More
Apexbio Technology LLC CFTRinh-172, 10mg. Cas: 307510-92-5 MFCD: MFCD01164609. CFTR inhibitor, highly potent and selective
SDP
Supplier:  Apexbio Technology LLC B143510
Encompass_Preferred
CFTRinh-172 is a voltage-independent, selective CFTR inhibitor with K i of 300 nM, showing no effects on MDR1, ATP-sensitive K+ channels, or a series of other transporters. 307510-92-5. MFCD01164609
Catalog No. 50-101-2049