Learn More
Apexbio Technology LLC CFTRinh-172, 50mg. Cas: 307510-92-5 MFCD: MFCD01164609. CFTR inhibitor, highly potent and selective
SDP
Catalog No. 501012050
Encompass_Preferred
CFTRinh-172 is a voltage-independent, selective CFTR inhibitor with K i of 300 nM, showing no effects on MDR1, ATP-sensitive K+ channels, or a series of other transporters. 307510-92-5. MFCD01164609
Catalog No. 50-101-2050 Supplier Apexbio Technology LLC Supplier No. B143550
CFTRinh-172 is a voltage-independent, selective CFTR inhibitor with K i of 300 nM, showing no effects on MDR1, ATP-sensitive K+ channels, or a series of other transporters. 307510-92-5. MFCD01164609