Learn More
Thermo Scientific Dionex™ IonPac™ CG 12 A IC Columns | | 1 EA
Catalog No. NC0408285 Shop All Thermo Scientific Products
Encompass
Separate common cations in as little as 4 minutes using MSA or sulfuric acid eluents and this carboxylate-functionalized cation-exchange column
Catalog No. NC0408285 Supplier Thermo Scientific Supplier No. 046076
Separate common cations in as little as 4 minutes using MSA or sulfuric acid eluents and this carboxylate-functionalized cation-exchange column