Learn More
Chemglass Life Sciences 500mL Bottle, Wide Neck, GLS 80 Thread, Graduated, Duran
SDP
Catalog No. NC1631094
Encompass_Preferred
500mL Bottle, Wide Neck, GLS 80 Thread, Graduated, Duran. Wide neck screw thread – 80 mm diameter, special thread design for rapid opening and closing. Inert and chemically resistant – exceptional properties of DURAN eliminate problems with ion leaching. Mechanical strength – the sturdy design of these wide neck bottles enhances safety and service life. Autoclavable – excellent thermal properties for autoclaving and dry sterilization. Transparency – contents and volume can be visually checked with ease. Traceability – the eight-digit batch code allows the bottles to be traced back to the point of production, including the matching batch. The end-user can use the corresponding item number to retrieve a batch and quality certificate for any DURAN laboratory bottle at any time via the Internet. Stability – the large, stable base area allows for safe and easy storage.
Catalog No. NC1631094 Supplier Chemglass Life Sciences Supplier No. CG81201
500mL Bottle, Wide Neck, GLS 80 Thread, Graduated, Duran. Wide neck screw thread – 80 mm diameter, special thread design for rapid opening and closing. Inert and chemically resistant – exceptional properties of DURAN eliminate problems with ion leaching. Mechanical strength – the sturdy design of these wide neck bottles enhances safety and service life. Autoclavable – excellent thermal properties for autoclaving and dry sterilization. Transparency – contents and volume can be visually checked with ease. Traceability – the eight-digit batch code allows the bottles to be traced back to the point of production, including the matching batch. The end-user can use the corresponding item number to retrieve a batch and quality certificate for any DURAN laboratory bottle at any time via the Internet. Stability – the large, stable base area allows for safe and easy storage.