Learn More
Chemglass Life Sciences Dish, Crystallizing, 2500mL, 190 x 100mm
SDP
Supplier:  Chemglass Life Sciences CG8276190
Encompass_Preferred
Dish, Crystallizing, 2500mL, 190 x 100mm. The rims on these dishes are reinforced and fire polished to reduce chipping. Will withstand repeated sterilization (wet or dry). Ideal for storage and crystallization.
Catalog No. NC1732445