Learn More
Thermo Scientific CG12A GUARD COL 4X50
Catalog No. NC9017323 Shop All Thermo Scientific Products
Encompass
IonPac CG12A-8 um Guard Column (4 x 50 mm), for Complex Samples, Maximum Operating Pressure:4000 psi, Mobile Phase Compatibility:Acidic Eluents, 100% Acetonitrile.Alcohols Should be Avoided, Bead Dia:8 um
Catalog No. NC9017323 Supplier Thermo Scientific Supplier No. 046074
IonPac CG12A-8 um Guard Column (4 x 50 mm), for Complex Samples, Maximum Operating Pressure:4000 psi, Mobile Phase Compatibility:Acidic Eluents, 100% Acetonitrile.Alcohols Should be Avoided, Bead Dia:8 um