Learn More
Sigma Aldrich Fine Chemicals Biosciences CGA 350 Gas Cylinder Press
Catalog No. 502908667
Encompass
For use with hydrogen, methane, or 5% methane in argon gas cylinders.
Catalog No. 50-290-8667 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 29591U
For use with hydrogen, methane, or 5% methane in argon gas cylinders.