Learn More
MATHESON TRI GAS CGA 580 CYLINDER CONNECTION
Catalog No. NC0918465
Encompass
CGA Fitting - w/Integral Check Valve (SS). CGA 580. These standard cylinder valve outlet connecters come with a 1/4" MNPT connection on the other end. A check valve is built into the tailpiece of the connection. Washers are included with CGA fittings that require washers. Stainless Steel/Viton.
Catalog No. NC0918465 Supplier MATHESON TRI GAS Supplier No. SEQ580CV4SS
CGA Fitting - w/Integral Check Valve (SS). CGA 580. These standard cylinder valve outlet connecters come with a 1/4" MNPT connection on the other end. A check valve is built into the tailpiece of the connection. Washers are included with CGA fittings that require washers. Stainless Steel/Viton.