Learn More
Aviva Systems Biology Corp CGA Antibody - N-terminal region (ARP51770_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51770_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CGA. It was validated on Western Blot. . Alternate names: CG-ALPHA, FSHA, GPHA1, GPHa, HCG, LHA, TSHAApplicationWB. Species reactivity: Human
Catalog No. 50-285-591