Learn More
AdipoGen cGAMP Disodium salt, 849214-04-6 (free acid), MFCD28411367, 500 ug
SDP
Supplier:  AdipoGen AGCR13588C500
Encompass_Preferred
MF: C20H24N10O13P2 . 2Na, MW: 674.4 . 46.0, ≥98%, Appearance: Crystalline solid. Potent STING ligand, second messenger in human and mouse cells that triggers interferon production in response to cytosolic DNA, binding to STING leads to activation of IRF3 (IFN regulatory factor 3) and the induction of interferon-β, first cyclic nucleotide in metazoa, and plays a role in bacterial chemotaxis and colonization
Catalog No. 50-596-578