Learn More
AdipoGen cGAMP 2Na STING Ligand
SDP
Catalog No. 502562272
Encompass_Preferred
Chemical. CAS 849214-04-6 free acid. Formula C20H24N10O13P2 . 2Na. MW 674.4 . 46.0. Synthetic. Potent STING ligand. Second messenger in human and mouse cells that triggers interferon production in response to cytosolic DNA. Binding to STING leads to activation of IRF3 IFN regulatory factor 3 and the induction of interferon-beta. First cyclic nucleotide in metazoa. Plays a role in bacterial chemotaxis and colonization.
Catalog No. 50-256-2272 Supplier AdipoGen Supplier No. AGCR13588C100
Chemical. CAS 849214-04-6 free acid. Formula C20H24N10O13P2 . 2Na. MW 674.4 . 46.0. Synthetic. Potent STING ligand. Second messenger in human and mouse cells that triggers interferon production in response to cytosolic DNA. Binding to STING leads to activation of IRF3 IFN regulatory factor 3 and the induction of interferon-beta. First cyclic nucleotide in metazoa. Plays a role in bacterial chemotaxis and colonization.