Learn More
Medchemexpress LLC cGAMP diammonium | 96.6% | 708.47 | 50 MG
SDP
Catalog No. 5000343223
Encompass_Preferred
cGAMP (Cyclic GMP-AMP) diammonium functions as an endogenous second messenger in metazoans and triggers interferon production in response to cytosolic DNA. It activates stimulator of interferon genes (STING), which activates a signaling cascade leading to the production of type I interferons and other immune mediators.
  • Endogenous second messenger in metazoans.
  • Triggers interferon production in response to cytosolic DNA.
  • Activates stimulator of interferon genes (STING).
  • Activates a signaling cascade leading to the production of type I interferons and other immune mediators.
  • Promotes the antigen-specific proliferation capacity of spleen cells in mice.
  • Directly activates murine and human dendritic cells *in vitro*.
Catalog No. 50-003-43223 Supplier Medchemexpress LLC Supplier No. HY110385A10MG
cGAMP (Cyclic GMP-AMP) diammonium functions as an endogenous second messenger in metazoans and triggers interferon production in response to cytosolic DNA. It activates stimulator of interferon genes (STING), which activates a signaling cascade leading to the production of type I interferons and other immune mediators.
  • Endogenous second messenger in metazoans.
  • Triggers interferon production in response to cytosolic DNA.
  • Activates stimulator of interferon genes (STING).
  • Activates a signaling cascade leading to the production of type I interferons and other immune mediators.
  • Promotes the antigen-specific proliferation capacity of spleen cells in mice.
  • Directly activates murine and human dendritic cells *in vitro*.