Learn More
Medchemexpress LLC cGAMP (diammonium) | 96.63% | 708.47 | 25 MG
SDP
Catalog No. 5000345807
Encompass_Preferred
cGAMP (Cyclic GMP-AMP) diammonium functions as an endogenous second messenger in metazoans and triggers interferon production in response to cytosolic DNA. It activates stimulator of interferon genes (STING), which leads to a signaling cascade that produces type I interferons and other immune mediators.
  • Functions naturally in metazoans
  • Responds to cytosolic DNA by triggering interferon production
  • Activates STING, initiating a signaling cascade for type I interferons and other immune mediators
  • Promotes the antigen-specific proliferation capacity of spleen cells in mice
  • Directly activates murine and human dendritic cells in vitro
  • Induces an antiviral state and secretion of type I IFNs
  • Promotes antigen-specific cytokine production by spleen cells of immunized mice
Catalog No. 50-003-45807 Supplier Medchemexpress LLC Supplier No. 25MGHY110385A
cGAMP (Cyclic GMP-AMP) diammonium functions as an endogenous second messenger in metazoans and triggers interferon production in response to cytosolic DNA. It activates stimulator of interferon genes (STING), which leads to a signaling cascade that produces type I interferons and other immune mediators.
  • Functions naturally in metazoans
  • Responds to cytosolic DNA by triggering interferon production
  • Activates STING, initiating a signaling cascade for type I interferons and other immune mediators
  • Promotes the antigen-specific proliferation capacity of spleen cells in mice
  • Directly activates murine and human dendritic cells in vitro
  • Induces an antiviral state and secretion of type I IFNs
  • Promotes antigen-specific cytokine production by spleen cells of immunized mice