Learn More
Cayman Chemical cGAS EnzymehumnrecombIna 1ea
SDP
Supplier:  Cayman Chemical 4005231 EA
Encompass_Preferred
cGAS Enzyme (human, recombinant) has been tested and formulated to work exclusively with Cayman’s cGAS TR-FRET Inhibitor Screening Assay Kit (Item No. 702120).
Catalog No. 50-294-2432