Learn More
Medchemexpress LLC cGAS Human His 100ug
SDP
Catalog No. 5000182962
Encompass_Preferred
cGAS is a cytosolic DNA sensor cGAS binds double-stranded DNA (dsDNA) and catalyzes the generation of cyclic guanosine-adenylate (cGAMP) which in turn activates the STING protein cGAS induces the production of type I interferon (IFN-I) and pro-inflammatory cytokines cGAS Protein Human (His) is a recombinant cGAS protein expressed in E coli with a C-6 His tag
Catalog No. 50-001-82962 Supplier Medchemexpress LLC Supplier No. HYP72337100UG
cGAS is a cytosolic DNA sensor cGAS binds double-stranded DNA (dsDNA) and catalyzes the generation of cyclic guanosine-adenylate (cGAMP) which in turn activates the STING protein cGAS induces the production of type I interferon (IFN-I) and pro-inflammatory cytokines cGAS Protein Human (His) is a recombinant cGAS protein expressed in E coli with a C-6 His tag