Learn More
Medchemexpress LLC cGAS Human His-SUM 100ug
SDP
Catalog No. 5000196698
Encompass_Preferred
Recombinant human cGAS (fragment G161-F522) expressed in E. coli with N-terminal 6xHis and SUMO tags. Supplied lyophilized with >90% purity and formulated in Tris-HCl or PBS with trehalose, this protein is suitable for biochemical and binding studies of cytosolic DNA sensing and cGAMP synthesis. Store lyophilized at -20°C; after reconstitution it is stable at 4°C for one week or at -20°C for extended storage when aliquoted.
Catalog No. 50-001-96698 Supplier Medchemexpress LLC Supplier No. HYP71597100UG
Recombinant human cGAS (fragment G161-F522) expressed in E. coli with N-terminal 6xHis and SUMO tags. Supplied lyophilized with >90% purity and formulated in Tris-HCl or PBS with trehalose, this protein is suitable for biochemical and binding studies of cytosolic DNA sensing and cGAMP synthesis. Store lyophilized at -20°C; after reconstitution it is stable at 4°C for one week or at -20°C for extended storage when aliquoted.