Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC cGAS Human His-SUM 10ug
SDP

Catalog No. 5000196328
Encompass_Preferred

Recombinant human cyclic GMP-AMP synthase (cGAS) fragment G161-F522, expressed in E. coli and supplied lyophilized with an N-terminal 6xHis-SUMO tag. The protein is purified to >90% and provided with storage and reconstitution guidance to preserve activity for biochemical and structural applications.

  • Recombinant human cGAS fragment G161-F522 with N-terminal His-SUMO tag.
  • Expressed in E. coli and purified to greater than 90% purity.
  • Lyophilized formulation for improved stability and convenient storage.
  • Buffer contains trehalose to protect protein during lyophilization.
  • Suitable for biochemical assays, enzymatic activity, and structural studies.
  • Includes storage and reconstitution guidance to maintain activity.

Catalog No. 50-001-96328 Supplier Medchemexpress LLC Supplier No. HYP7159710UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human cyclic GMP-AMP synthase (cGAS) fragment G161-F522, expressed in E. coli and supplied lyophilized with an N-terminal 6xHis-SUMO tag. The protein is purified to >90% and provided with storage and reconstitution guidance to preserve activity for biochemical and structural applications.

  • Recombinant human cGAS fragment G161-F522 with N-terminal His-SUMO tag.
  • Expressed in E. coli and purified to greater than 90% purity.
  • Lyophilized formulation for improved stability and convenient storage.
  • Buffer contains trehalose to protect protein during lyophilization.
  • Suitable for biochemical assays, enzymatic activity, and structural studies.
  • Includes storage and reconstitution guidance to maintain activity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.