Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC cGAS Human His-SUM 50ug
SDP

Catalog No. 5000196697
Encompass_Preferred

Recombinant human cyclic GMP-AMP synthase (cGAS) fragment (G161-F522) expressed in E. coli and supplied lyophilized with N-terminal 6xHis and SUMO tags to aid purification and solubility. Supplied amount for this item is 50 μg; other pack sizes are available. Activity testing is in progress and not guaranteed. Store lyophilized material frozen and aliquot after reconstitution per manufacturer's guidance.

  • Recombinant fragment spanning residues G161-F522.
  • N-terminal 6xHis and SUMO tags for purification and solubility.
  • Expressed in E. coli for bacterial expression.
  • Lyophilized formulation with trehalose for improved stability.
  • Available in multiple pack sizes, including 50 μg.
  • Activity testing in progress; no guaranteed activity.
  • Recommend aliquoting and storage at -20°C or -80°C for long-term preservation.

Catalog No. 50-001-96697 Supplier Medchemexpress LLC Supplier No. HYP7159750UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human cyclic GMP-AMP synthase (cGAS) fragment (G161-F522) expressed in E. coli and supplied lyophilized with N-terminal 6xHis and SUMO tags to aid purification and solubility. Supplied amount for this item is 50 μg; other pack sizes are available. Activity testing is in progress and not guaranteed. Store lyophilized material frozen and aliquot after reconstitution per manufacturer's guidance.

  • Recombinant fragment spanning residues G161-F522.
  • N-terminal 6xHis and SUMO tags for purification and solubility.
  • Expressed in E. coli for bacterial expression.
  • Lyophilized formulation with trehalose for improved stability.
  • Available in multiple pack sizes, including 50 μg.
  • Activity testing in progress; no guaranteed activity.
  • Recommend aliquoting and storage at -20°C or -80°C for long-term preservation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.