Learn More
Medchemexpress LLC cGAS Human His-SUM 5ug
SDP
Catalog No. 5000196327
Encompass_Preferred
The cGAS protein is a nucleotidyl transferase that crucially mediates innate immunity by catalyzing the formation of 2 3 -cGAMP from ATP and GTP upon binding to double-stranded DNA (dsDNA) As a DNA sensor cGAS detects exogenous and endogenous DNA triggering STING1 activation and type I interferon production cGAS Protein Human (His-SUMO) is the recombinant human-derived cGAS protein expressed by E coli with N-6 His N-SUMO labeled tag
Catalog No. 50-001-96327 Supplier Medchemexpress LLC Supplier No. HYP715975UG
The cGAS protein is a nucleotidyl transferase that crucially mediates innate immunity by catalyzing the formation of 2 3 -cGAMP from ATP and GTP upon binding to double-stranded DNA (dsDNA) As a DNA sensor cGAS detects exogenous and endogenous DNA triggering STING1 activation and type I interferon production cGAS Protein Human (His-SUMO) is the recombinant human-derived cGAS protein expressed by E coli with N-6 His N-SUMO labeled tag