Learn More
Cayman Chemical cGAS (human recombinant), 100 µg
SDP
Supplier:  Cayman Chemical 22810100
Encompass_Preferred
Source: N-terminally Histidine-tagged human cGAS protein, purified from E. coli • Amino acids: 2-543 (full length) • MW: 61 kDa
Catalog No. NC1486600