Learn More
Cayman Chemical cGAS MoncLNal ANTIBDYClo 300ug
SDP
Catalog No. 502995266
Encompass_Preferred
Immunogen: Full length human recombinant protein • Host: Mouse, clone 5G10 • Species Reactivity: (+) Human cGAS • Application(s): WB, IF, and IP
Catalog No. 50-299-5266 Supplier Cayman Chemical Supplier No. 23853300 UG
Immunogen: Full length human recombinant protein • Host: Mouse, clone 5G10 • Species Reactivity: (+) Human cGAS • Application(s): WB, IF, and IP