Learn More
Cayman Chemical cGAS ReactIon Buffer2X 6mL
SDP
Catalog No. 502896634
Encompass_Preferred
cGAS Reaction Buffer (2X) has been tested and formulated to work exclusively with Cayman’s cGAS Inhibitor Screening Assay Kit (Item No. 701930).
Catalog No. 50-289-6634 Supplier Cayman Chemical Supplier No. 7019316 ML
cGAS Reaction Buffer (2X) has been tested and formulated to work exclusively with Cayman’s cGAS Inhibitor Screening Assay Kit (Item No. 701930).