Learn More
Cayman Chemical cGAS Stop Solution, 1 mL
SDP
Catalog No. 502896703
Encompass_Preferred
cGAS Stop Solution has been tested and formulated to work exclusively with Cayman’s cGAS Inhibitor Screening Assay Kit (Item No. 701930).
Catalog No. 50-289-6703 Supplier Cayman Chemical Supplier No. 7019351 ML
cGAS Stop Solution has been tested and formulated to work exclusively with Cayman’s cGAS Inhibitor Screening Assay Kit (Item No. 701930).