Learn More
Cayman Chemical cGAS TR-FRET Stop SolutIon 1ea
SDP
Catalog No. 502942978
Encompass_Preferred
cGAS TR-FRET Stop Solution has been tested and formulated to work exclusively with Cayman’s cGAS TR-FRET Inhibitor Screening Assay Kit (Item No. 702120).
Catalog No. 50-294-2978 Supplier Cayman Chemical Supplier No. 4005211 EA
cGAS TR-FRET Stop Solution has been tested and formulated to work exclusively with Cayman’s cGAS TR-FRET Inhibitor Screening Assay Kit (Item No. 702120).