Learn More
Cayman Chemical cGAS161-522humn recombI 100ug
SDP
Catalog No. 502933914
Encompass_Preferred
Source: N-terminal His-tagged human cGAS protein (truncated), purified from E. coli • Amino acids: 161-522 • MW: 44.6 kDa
Catalog No. 50-293-3914 Supplier Cayman Chemical Supplier No. 25001100 UG
Source: N-terminal His-tagged human cGAS protein (truncated), purified from E. coli • Amino acids: 161-522 • MW: 44.6 kDa