Learn More
Aviva Systems Biology Corp CGB3 Antibody - C-terminal region (ARP60118_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60118_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CGB. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CGB. Alternate names: CGB3, CGB5, CGB7, CGB8, hCGBApplicationWB. Species reactivity: Cow, Human, Rabbit
Catalog No. 50-286-946