Learn More
Sino Biological CGB5 / CG-beta Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 14574T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CGB5 / CG-beta (rh CGB5 / CG-beta; Catalog#14574-H08H; P01233-1; Met1-Gln165). CGB5 / CG-beta specific IgG was purified by Human CGB5 / CG-beta affinity chromatography.
Catalog No. 50-224-7472